Learn More
AdipoGen Limonin
SDP
Catalog No. 502560599
Encompass_Preferred
Chemical. CAS 1180-71-8. Formula C26H30O8. MW 470.51. Synthetic. Limonin is the first described limonoid, which are naturally occurring tetranortriterpenoids that impart bitter flavor in citrus fruits. Compounds belonging to this group have reported anti-proliferative and proapoptotic activity on several cancer cell lines as well as pesticidal, anti-malarial, anti-microbial, anti-HIV, anti-inflammatory, antinociceptive and neuroprotective activity. Shown to inhibit arginase I and II and P-glycoprotein. May be used in assays as control, standard or competitor, or in studies on the physiology, metabolism and biosynthesis of limonin.
Catalog No. 50-256-0599 Supplier AdipoGen Supplier No. CDXL0280M050
Chemical. CAS 1180-71-8. Formula C26H30O8. MW 470.51. Synthetic. Limonin is the first described limonoid, which are naturally occurring tetranortriterpenoids that impart bitter flavor in citrus fruits. Compounds belonging to this group have reported anti-proliferative and proapoptotic activity on several cancer cell lines as well as pesticidal, anti-malarial, anti-microbial, anti-HIV, anti-inflammatory, antinociceptive and neuroprotective activity. Shown to inhibit arginase I and II and P-glycoprotein. May be used in assays as control, standard or competitor, or in studies on the physiology, metabolism and biosynthesis of limonin.