Learn More
Cayman Chemical Limonin 17-β-D-Glucopyranoside, Purity: ≥95%, CAS: 123564-61-4, 1 mg
SDP
Catalog No. 502918676
Encompass_Preferred
A furanolactone with anticancer activity; induces apoptosis and disrupts the mitochondrial membrane potential in SW480 cells at 100 µM; inhibits the proliferation of MDA-Panc-28 cells (IC50 = 20.49 µM)
Catalog No. 50-291-8676 Supplier Cayman Chemical Supplier No. 391471 MG
A furanolactone with anticancer activity; induces apoptosis and disrupts the mitochondrial membrane potential in SW480 cells at 100 µM; inhibits the proliferation of MDA-Panc-28 cells (IC50 = 20.49 µM)