Learn More
Enzo Life Sciences Limonin (50mg). CAS: 1180-71-8
SDP
Supplier:  Enzo Life Sciences ALX350225M050
Encompass_Preferred
Bitter principle of citrus fruits. Inhibits chemically induced carcinogenesis. Inhibits HIV-1 replication. Shows antinociceptive effect. Does not possess any inherent antioxidant activity. Alternative name: Limonoic acid di-delta-lactone. Purity: ≥98% (UHPLC). Appearance: White to off-white solid. Solubility: Soluble in 100% ethanol or glacial acetic acid; slightly soluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-8786