Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Limonin, >99.6%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN19376505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19376505 5 mg
MP219376510 10 mg
MP219376525 25 mg
MP219376550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19376505 Supplier MP Biomedicals, Inc Supplier No. 0219376505
Add to Cart
Add to Cart

Limonin from Citrus Seeds White Crystalline Powder

  • It may be used in assays to quantify limonin in citrus products.
  • It may also be used in other assays as a control, standard, or competitor or in studies on the physiology, metabolism, and biosynthesis of limonin.
  • It is a natural compound found in grapefruit and other citrus seeds.
  • IT is a triterpenoid aglycone that is a bitter principle of citrus fruits.
  • It has been shown to inhibit chemically induced carcinogenesis and demonstrates anti feedant properties.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1180-71-8
Molecular Formula C26H30O8
Molecular Weight (g/mol) 470.52
MDL Number MFCD00075922
InChI Key KBDSLGBFQAGHBE-MSGMIQHVSA-N
PubChem CID 179651
ChEBI CHEBI:16226
IUPAC Name (1R,2R,7S,10R,13R,14R,16S,19S,20S)-19-(furan-3-yl)-9,9,13,20-tetramethyl-4,8,15,18-tetraoxahexacyclo[11.9.0.0²,⁷.0²,¹⁰.0¹⁴,¹⁶.0¹⁴,²⁰]docosane-5,12,17-trione
SMILES CC1(C)O[C@H]2CC(=O)OC[C@@]22[C@H]1CC(=O)[C@]1(C)[C@@H]2CC[C@@]2(C)[C@@H](OC(=O)[C@H]3O[C@@]123)C1=COC=C1

Specifications

Color White
Assay Percent Range >99.6%
Quantity 5 mg
Synonym limonin, dictamnolactone, obaculactone, citrolimonin, evodin, 7,16-dioxo-7,16-dideoxylimondiol, limoni, limonoic acid, di-delta-lactone, limonine, limonoate d-ring-lactone
Formula Weight 470.5
Physical Form Powder
Chemical Name or Material Limonin, For HPLC analysis
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.