Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- It may be used in assays to quantify limonin in citrus products.
- It may also be used in other assays as a control, standard, or competitor or in studies on the physiology, metabolism, and biosynthesis of limonin.
- It is a natural compound found in grapefruit and other citrus seeds.
- IT is a triterpenoid aglycone that is a bitter principle of citrus fruits.
- It has been shown to inhibit chemically induced carcinogenesis and demonstrates anti feedant properties.
Specifications
Specifications
| Melting Point | 298°C to 300°C (literature) |
| Color | White |
| Loss on Drying | 0.0038 |
| Quantity | 25 mg |
| Synonym | Limonoic acid 3, 19:16,17 dilactone |
| Solubility Information | Soluble in water, ether; soluble in alcohol,glacial acetic acid. |
| Physical Form | Crystalline Powder |
| Grade | 0.999 |
Safety and Handling
| storageNote1 | Store at+4°C. |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?