Learn More
Medchemexpress LLC Limonin (Standard) | 1180-71-8 | 99.1% | 470.51 | 25 MG
SDP
Catalog No. 5000384018
Encompass_Preferred
Limonin (Standard) is an analytical and reference standard intended for research and analytical applications. It inhibits HIV-1 with an EC50 of 60.0 μM and induces human colon adenocarcinoma cells apoptosis with an IC50 of 54.74 μM, possessing antiviral and antitumor activities.
  • Intended for research and analytical applications.
  • Inhibits HIV-1 with an EC50 of 60.0 μM.
  • Induces human colon adenocarcinoma cells apoptosis with an IC50 of 54.74 μM.
  • Possesses antiviral and antitumor activities.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-84018 Supplier Medchemexpress LLC Supplier No. 25MGHY17411R
Limonin (Standard) is an analytical and reference standard intended for research and analytical applications. It inhibits HIV-1 with an EC50 of 60.0 μM and induces human colon adenocarcinoma cells apoptosis with an IC50 of 54.74 μM, possessing antiviral and antitumor activities.
  • Intended for research and analytical applications.
  • Inhibits HIV-1 with an EC50 of 60.0 μM.
  • Induces human colon adenocarcinoma cells apoptosis with an IC50 of 54.74 μM.
  • Possesses antiviral and antitumor activities.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.