Learn More
Medchemexpress LLC Limonin (Standard) | 1180-71-8 | MFC26H30O8 | 99.1% | 470.51 | 50 MG
SDP
Catalog No. 5000385297
Encompass_Preferred
Limonin (Standard) is an analytical standard used for research and analytical applications. It exhibits antiviral and antitumor activities, including the inhibition of HIV-1 and induction of apoptosis in human colon adenocarcinoma cells. This standard is commonly employed in qualitative, quantitative, and methodological research experiments.
  • Inhibits HIV-1 with an EC50 of 60.0 μM.
  • Induces apoptosis in human colon adenocarcinoma cells with an IC50 of 54.74 μM.
  • Used in HPLC, GC, and MS applications.
  • Appears as a white to off-white solid.
  • Classified as a terpenoid and triterpene.
  • Initially sourced from plants of the Rutaceae family.
  • Functions as an endogenous metabolite.
Catalog No. 50-003-85297 Supplier Medchemexpress LLC Supplier No. HY17411R50MG
Limonin (Standard) is an analytical standard used for research and analytical applications. It exhibits antiviral and antitumor activities, including the inhibition of HIV-1 and induction of apoptosis in human colon adenocarcinoma cells. This standard is commonly employed in qualitative, quantitative, and methodological research experiments.
  • Inhibits HIV-1 with an EC50 of 60.0 μM.
  • Induces apoptosis in human colon adenocarcinoma cells with an IC50 of 54.74 μM.
  • Used in HPLC, GC, and MS applications.
  • Appears as a white to off-white solid.
  • Classified as a terpenoid and triterpene.
  • Initially sourced from plants of the Rutaceae family.
  • Functions as an endogenous metabolite.