Learn More
Medchemexpress LLC Limonin (standard) | 1180-71-8 | MFCD00075922 | 99.1% | 470.51 | 5 MG
SDP
Catalog No. 5000384602
Encompass_Preferred
Limonin (Standard) is the analytical standard of Limonin, intended for research and analytical applications. It exhibits antiviral and antitumor activities, inhibiting HIV-1 with an EC50 of 60.0 μM and inducing apoptosis in human colon adenocarcinoma cells with an IC50 of 54.74 μM. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Inhibits HIV-1 and induces human colon adenocarcinoma cell apoptosis
  • Possesses antiviral and antitumor activities
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for applications such as HPLC, GC, and MS
Catalog No. 50-003-84602 Supplier Medchemexpress LLC Supplier No. HY17411R5MG
Limonin (Standard) is the analytical standard of Limonin, intended for research and analytical applications. It exhibits antiviral and antitumor activities, inhibiting HIV-1 with an EC50 of 60.0 μM and inducing apoptosis in human colon adenocarcinoma cells with an IC50 of 54.74 μM. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Inhibits HIV-1 and induces human colon adenocarcinoma cell apoptosis
  • Possesses antiviral and antitumor activities
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for applications such as HPLC, GC, and MS