Learn More
Cederlane LIMPHOLYTE MAMMAL 500ML
SDP
Catalog No. NC9035801
Encompass
Lympholyte-Mammal is a density separation medium specifically designed for the isolation of viable lymphocytes and monocytes from the peripheral blood of most mammalian species. It consists of Sodium Diatrizoate combined with Dextran to induce erythrocyte aggregation and reduce platelet aggregation resulting in a higher yield of lymphocytes and monocytes. Lympholyte-Mammal can be utilized with a simple protocol for the elimination of erythrocytes and dead cells from the blood of most mammalian species. Lympholyte-Mammal also removes the majority of granulocytes (including neutrophils). The resulting cell population demonstrates a high and non-selective recovery of viable lymphocytes and monocytes.
Catalog No. NC9035801 Supplier Cederlane Supplier No. CL5120
Lympholyte-Mammal is a density separation medium specifically designed for the isolation of viable lymphocytes and monocytes from the peripheral blood of most mammalian species. It consists of Sodium Diatrizoate combined with Dextran to induce erythrocyte aggregation and reduce platelet aggregation resulting in a higher yield of lymphocytes and monocytes. Lympholyte-Mammal can be utilized with a simple protocol for the elimination of erythrocytes and dead cells from the blood of most mammalian species. Lympholyte-Mammal also removes the majority of granulocytes (including neutrophils). The resulting cell population demonstrates a high and non-selective recovery of viable lymphocytes and monocytes.