Learn More
Aviva Systems Biology Corp LIN28 Antibody - N-terminal region (ARP34477_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34477_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIN28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LIN28. Alternate names: CSDD1, FLJ12457, LIN-28, LIN28A, ZCCHC1, LIN28, lin-28AApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-55410