Learn More
Medchemexpress LLC Lin28 inhibitor LI71 enantiomer | 956189-58-5 | 99.7% | 335.40 g·mol⁻1 | C21H21NO3 | 5MG
SDP
Catalog No. 5000235754
Encompass_Preferred
LIN28 inhibitor LI71 enantiomer is the less active stereoisomer of a small-molecule LIN28 inhibitor used for research on LIN28-driven cancer biology and RNA-binding protein function. It disrupts LIN28:let-7 binding (reported IC50 ≈ 7 μM) and abolishes LIN28-mediated oligouridylation of the let-7 precursor (reported IC50 ≈ 27 μM); it has also been reported to affect parasite cold shock protein interactions with DNA and tubulin.
  • High purity suitable for research applications
  • Cell-permeable small molecule for cellular assays
  • Disrupts LIN28:let-7 binding (IC50 ≈ 7 μM)
  • Abolishes LIN28-mediated oligouridylation of let-7 precursor (IC50 ≈ 27 μM)
  • Offered in multiple milligram pack sizes for experimental flexibility
Catalog No. 50-002-35754 Supplier Medchemexpress LLC Supplier No. HY123905A5MG
LIN28 inhibitor LI71 enantiomer is the less active stereoisomer of a small-molecule LIN28 inhibitor used for research on LIN28-driven cancer biology and RNA-binding protein function. It disrupts LIN28:let-7 binding (reported IC50 ≈ 7 μM) and abolishes LIN28-mediated oligouridylation of the let-7 precursor (reported IC50 ≈ 27 μM); it has also been reported to affect parasite cold shock protein interactions with DNA and tubulin.
  • High purity suitable for research applications
  • Cell-permeable small molecule for cellular assays
  • Disrupts LIN28:let-7 binding (IC50 ≈ 7 μM)
  • Abolishes LIN28-mediated oligouridylation of let-7 precursor (IC50 ≈ 27 μM)
  • Offered in multiple milligram pack sizes for experimental flexibility