Learn More
Enzo Life Sciences Lin28a (full-length) (human), (recombinant) (200µg)
SDP
Catalog No. 502013961
Encompass_Preferred
Alternative name: Protein lin-28 homolog A. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 20mM TRIS, pH 7.2, containing 200mM sodium chloride and 5mM diothiothreitol (DTT). Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-201-3961 Supplier Enzo Life Sciences Supplier No. ENZPRT1720200
Alternative name: Protein lin-28 homolog A. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 20mM TRIS, pH 7.2, containing 200mM sodium chloride and 5mM diothiothreitol (DTT). Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .