Learn More
ABclonal Technology LIN28A Rabbit PolymAb
SDP
Catalog No. 5000006575
Encompass_Preferred
This gene encodes a LIN-28 family RNA-binding protein that acts as a posttranscriptional regulator of genes involved in developmental timing and self-renewal in embryonic stem cells The encoded protein functions through direct interaction with target mRNAs and by disrupting the maturation of certain miRNAs involved in embryonic development This protein prevents the terminal processing of the LET7 family of microRNAs which are major regulators of cellular growth and differentiation Aberrant expression of this gene is associated with cancer progression in multiple tissues
Catalog No. 50-000-06575 Supplier ABclonal Technology Supplier No. A22648PM100UL
This gene encodes a LIN-28 family RNA-binding protein that acts as a posttranscriptional regulator of genes involved in developmental timing and self-renewal in embryonic stem cells The encoded protein functions through direct interaction with target mRNAs and by disrupting the maturation of certain miRNAs involved in embryonic development This protein prevents the terminal processing of the LET7 family of microRNAs which are major regulators of cellular growth and differentiation Aberrant expression of this gene is associated with cancer progression in multiple tissues