Learn More
Aviva Systems Biology Corp LIN37 Antibody - middle region (ARP57335_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57335_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIN37. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LIN37. Alternate names: F25965, MGC9505, ZK418.4, lin-37ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62126