Learn More
Aviva Systems Biology Corp LIN9 Antibody - N-terminal region (ARP50821_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50821_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LIN9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LIN9. Alternate names: TGS, BARA, TGS1, Lin-9, BARPsvApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53641