Learn More
Cayman Chemical Linaclotide, Purity: ≥95%, CAS: 851199-59-2, 10 mg
SDP
Catalog No. 502933149
Encompass_Preferred
A peptide agonist of the guanylate cyclase C receptor (Ki = 16.4 nM in a radioligand binding assay using mouse intestinal mucosa); stimulates fluid secretion and cGMP concentration in jejunal loops isolated from wild-type mice but not guanylate cyclase C receptor-null mice; increases intestinal transit rate in wild-type mice when administered at a dose of 100 μg/kg; reduces the number of phosphorylated ERK-positive dorsal horn neurons in the thoracolumbar spinal cord, a marker of nociceptive signaling, following noxious colorectal distension and mechanical hypersensitivity in a mouse model of TNBS-induced colitis
Catalog No. 50-293-3149 Supplier Cayman Chemical Supplier No. 2408510 MG
A peptide agonist of the guanylate cyclase C receptor (Ki = 16.4 nM in a radioligand binding assay using mouse intestinal mucosa); stimulates fluid secretion and cGMP concentration in jejunal loops isolated from wild-type mice but not guanylate cyclase C receptor-null mice; increases intestinal transit rate in wild-type mice when administered at a dose of 100 μg/kg; reduces the number of phosphorylated ERK-positive dorsal horn neurons in the thoracolumbar spinal cord, a marker of nociceptive signaling, following noxious colorectal distension and mechanical hypersensitivity in a mouse model of TNBS-induced colitis