Learn More
Medchemexpress LLC Linaclotide (acetate) | 851199-60-5 | 98.6% | 1586.79 | 25 MG
SDP
Catalog No. 5000384291
Encompass_Preferred
Linaclotide acetate is a potent and selective guanylate cyclase C agonist, developed for the treatment of constipation-predominant irritable bowel syndrome (IBS-C) and chronic constipation. This product is for research use only.
  • Treats constipation-predominant irritable bowel syndrome (IBS-C).
  • Treats chronic constipation.
Catalog No. 50-003-84291 Supplier Medchemexpress LLC Supplier No. 25MGHY17584A
Linaclotide acetate is a potent and selective guanylate cyclase C agonist, developed for the treatment of constipation-predominant irritable bowel syndrome (IBS-C) and chronic constipation. This product is for research use only.
  • Treats constipation-predominant irritable bowel syndrome (IBS-C).
  • Treats chronic constipation.