Learn More
Medchemexpress LLC Linaclotide (acetate) | 851199-60-5 | 98.6% | 1586.79 | 5 MG
SDP
Catalog No. 5000387469
Encompass_Preferred
Linaclotide acetate is a potent and selective guanylate cyclase C agonist, developed to address conditions such as constipation-predominant irritable bowel syndrome (IBS-C) and chronic constipation. It functions by activating guanylate cyclase C, which helps to regulate fluid secretion and gastrointestinal transit. This compound is intended for research purposes only and is not sold to patients.
  • Potent and selective guanylate cyclase C agonist.
  • Developed for treating constipation-predominant irritable bowel syndrome.
  • Developed for treating chronic constipation.
  • Available in various quantities for research needs.
  • High purity of 98.61% ensures reliable experimental results.
  • Detailed documentation including data sheet, COA, and SDS is provided.
  • Solubility information and preparation protocols are available for in vitro and in vivo studies.
  • Supported by scientific publications.
Catalog No. 50-003-87469 Supplier Medchemexpress LLC Supplier No. HY17584A5MG
Linaclotide acetate is a potent and selective guanylate cyclase C agonist, developed to address conditions such as constipation-predominant irritable bowel syndrome (IBS-C) and chronic constipation. It functions by activating guanylate cyclase C, which helps to regulate fluid secretion and gastrointestinal transit. This compound is intended for research purposes only and is not sold to patients.
  • Potent and selective guanylate cyclase C agonist.
  • Developed for treating constipation-predominant irritable bowel syndrome.
  • Developed for treating chronic constipation.
  • Available in various quantities for research needs.
  • High purity of 98.61% ensures reliable experimental results.
  • Detailed documentation including data sheet, COA, and SDS is provided.
  • Solubility information and preparation protocols are available for in vitro and in vivo studies.
  • Supported by scientific publications.