Learn More
Medchemexpress LLC Linaclotide acetate | 851199-60-5 | 98.6% | 1586.79 | C61H83N15O23S6 | 10 MG
SDP
Catalog No. 5000298703
Encompass_Preferred
Linaclotide acetate is a potent, selective guanylate cyclase C (GC-C) agonist developed for treatment of constipation-predominant irritable bowel syndrome (IBS-C) and chronic constipation. Supplied as a research-grade peptide, it acts on intestinal GC-C receptors to increase chloride and bicarbonate secretion and accelerate intestinal transit, supporting pharmacological and translational studies under appropriate laboratory conditions.
  • Potent, selective guanylate cyclase C agonist
  • Promotes chloride and bicarbonate secretion to increase intestinal fluid
  • Peptide suitable for research-grade in vitro and in vivo studies
  • High purity and characterized molecular weight for reproducible results
  • Stable when stored frozen; follow recommended storage conditions
Catalog No. 50-002-98703 Supplier Medchemexpress LLC Supplier No. HY17584A10MG
Linaclotide acetate is a potent, selective guanylate cyclase C (GC-C) agonist developed for treatment of constipation-predominant irritable bowel syndrome (IBS-C) and chronic constipation. Supplied as a research-grade peptide, it acts on intestinal GC-C receptors to increase chloride and bicarbonate secretion and accelerate intestinal transit, supporting pharmacological and translational studies under appropriate laboratory conditions.
  • Potent, selective guanylate cyclase C agonist
  • Promotes chloride and bicarbonate secretion to increase intestinal fluid
  • Peptide suitable for research-grade in vitro and in vivo studies
  • High purity and characterized molecular weight for reproducible results
  • Stable when stored frozen; follow recommended storage conditions