Learn More
Medchemexpress LLC HY-10284 100mg Medchemexpress, Linagliptin CAS:668270-12-0 Purity:>98%
SDP
Catalog No. 501967999
Encompass_Preferred
Medchemexpress, HY-10284 100mg Linagliptin CAS:668270-12-0 Linagliptin is a highly potent, selective DPP-4 inhibitor with IC50 of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7999 Supplier Medchemexpress LLC Supplier No. HY10284100MG
Add to Cart
Add to Cart
Medchemexpress, HY-10284 100mg Linagliptin CAS:668270-12-0 Linagliptin is a highly potent, selective DPP-4 inhibitor with IC50 of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.