Learn More
AdipoGen Linagliptin (50 mg)
SDP
Supplier:  AdipoGen AGCR13618M050
Encompass_Preferred
Chemical. CAS: 668270-12-0. Formula: C25H28N8O2. MW: 472.5. Antidiabetic agent. Highly potent and selective competitive inhibitor of dipeptidyl-peptidase 4 (DPP4; DPP IV; CD26), an enzyme that degrades, glucagon-like peptide-1 (GLP-1) and glucose-dependent insulinotropic polypeptide (GIP). Prevents the inactivation of endogenous GLP-1 and GIP. Shown to restore beta cell function and survival in human isolated islets through GLP-1 stabilization. Improves insulin sensitivity. Anti-inflammatory compound.
Catalog No. 50-205-3986