Learn More
Selleck Chemical LLC Linagliptin 5mg 668270-12-0 BI-1356
SDP
Catalog No. 501363281
Encompass_Preferred
Linagliptin (BI-1356) is a highly potent, selective DPP-4 inhibitor with IC50 of 1 nM and exhibits a 10,000-fold higher selectivity for DPP-4 than for other dipeptidyl peptidases such as DPP-2, DPP-8, and DPP-9. Linagliptin activates glomerular autophagy in a model of type 2 diabetes. DPP4 mediates ferroptosis in TP53-deficient CRC cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3281 Supplier Selleck Chemical LLC Supplier No. S30315MG
Linagliptin (BI-1356) is a highly potent, selective DPP-4 inhibitor with IC50 of 1 nM and exhibits a 10,000-fold higher selectivity for DPP-4 than for other dipeptidyl peptidases such as DPP-2, DPP-8, and DPP-9. Linagliptin activates glomerular autophagy in a model of type 2 diabetes. DPP4 mediates ferroptosis in TP53-deficient CRC cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.