Learn More
Apexbio Technology LLC Linagliptin (BI-1356) 668270-12-0 10mg
SDP
Catalog No. 5000095417
Encompass_Preferred
Linagliptin (BI-1356 CAS 668270-12-0) is a potent competitive inhibitor of dipeptidyl peptidase-4 (DPP-4) demonstrating IC50 values of 0 4 1 1 nM in multiple in vitro assays using DPP-4 extracted from Caco-2 cells DPP-4 is a membrane-bound and soluble protease involved in the degradation of incretin hormones such as GLP-1 which enhance pancreatic -cell glucose sensitivity and insulin secretion Linagliptin displays marked selectivity for DPP-4 over related peptidases and serine proteases In vivo linagliptin dose-dependently inhibits plasma DPP-4 activity and sustains glycemic control with prolonged effects observed at pharmacologically relevant doses This compound is widely utilized in studies of glucose metabolism and -cell function
Catalog No. 50-000-95417 Supplier Apexbio Technology LLC Supplier No. A403410
Linagliptin (BI-1356 CAS 668270-12-0) is a potent competitive inhibitor of dipeptidyl peptidase-4 (DPP-4) demonstrating IC50 values of 0 4 1 1 nM in multiple in vitro assays using DPP-4 extracted from Caco-2 cells DPP-4 is a membrane-bound and soluble protease involved in the degradation of incretin hormones such as GLP-1 which enhance pancreatic -cell glucose sensitivity and insulin secretion Linagliptin displays marked selectivity for DPP-4 over related peptidases and serine proteases In vivo linagliptin dose-dependently inhibits plasma DPP-4 activity and sustains glycemic control with prolonged effects observed at pharmacologically relevant doses This compound is widely utilized in studies of glucose metabolism and -cell function