Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linagliptin-d4 | 2140263-92-7 | 98.2% | 476.57 g/mol | C25H24D4N8O2 | 5 MG
SDP

Catalog No. 5000245759
Encompass_Preferred

Linagliptin-d4 is a deuterium-labeled analogue of linagliptin intended for use as a tracer and internal standard in quantitative analytical workflows. It is suitable for NMR, GC-MS, and LC-MS quantitation, and may be used in pharmacokinetic and metabolic studies; the molecule also contains an alkyne moiety compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC).

  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Contains an alkyne moiety compatible with CuAAC click chemistry.
  • High listed purity (98.19%).
  • Molecular weight 476.57 g/mol for accurate mass calculations.
  • Provided in a 5 mg quantity for analytical and tracer studies.

Catalog No. 50-002-45759 Supplier Medchemexpress LLC Supplier No. HY10284S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linagliptin-d4 is a deuterium-labeled analogue of linagliptin intended for use as a tracer and internal standard in quantitative analytical workflows. It is suitable for NMR, GC-MS, and LC-MS quantitation, and may be used in pharmacokinetic and metabolic studies; the molecule also contains an alkyne moiety compatible with copper-catalyzed azide-alkyne cycloaddition (CuAAC).

  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Contains an alkyne moiety compatible with CuAAC click chemistry.
  • High listed purity (98.19%).
  • Molecular weight 476.57 g/mol for accurate mass calculations.
  • Provided in a 5 mg quantity for analytical and tracer studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.