Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Linalool

Catalog No. 62139100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-139-100ML 100 mL
62-139-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 62-139-100ML Supplier Sigma Aldrich Supplier No. 62139100ML
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-Linalool is a monoterpene compound mainly found in many essential oils. It shows anti-nociceptive and anti-inflammatory activity.(-)-Linalool can be used as a reactant in: The enantioselective preparation of ophiobolin sesterterpene via diastereoselective reductive radical cascade cyclization. The total synthesis of (-)-5,6-dihydrocineromycin B via (-)-linalool O-triethylsilyl ether.; The total synthesis of pladienolide B analog.Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 62139.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Specifications

CAS 126-91-0
Density 0.862 g/mL at 20°C
Boiling Point 198°C
Molecular Formula C10H18O
Refractive Index n20/D 1.4615; n20/D 1.462
Linear Formula (CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
MDL Number MFCD00135469
Quantity 100 mL
Synonym (R)-(-)-3,7-Dimethyl-1,6-octadien-3-ol
Molecular Weight (g/mol) 154.25 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.