Learn More
Selleck Chemical LLC Linalool S4957-25mg
SDP
Catalog No. 503109060
Encompass_Preferred
Linalool (Phantol) a monoterpene compound commonly found as major component of essential oils of several aromatic species is a competitive antagonist of NMDA receptors
Catalog No. 50-310-9060 Supplier Selleck Chemical LLC Supplier No. S495725MG
Linalool (Phantol) a monoterpene compound commonly found as major component of essential oils of several aromatic species is a competitive antagonist of NMDA receptors