Learn More
Medchemexpress LLC Linalool | 78-70-6 | 154.25 | 100 G
SDP
Catalog No. 5000389173
Encompass_Preferred
Linalool is a natural monoterpene that acts as a competitive NMDA receptor antagonist. It is orally active and can cross the blood-brain barrier.
  • Anticancer activity
  • Antibacterial properties
  • Anti-inflammatory effects
  • Neuroprotective qualities
  • Anxiolytic effects
  • Antidepressant properties
  • Anti-stress effects
  • Cardioprotective benefits
  • Hepatoprotective effects
  • Nephroprotective effects
  • Pulmonary protective activities
Catalog No. 50-003-89173 Supplier Medchemexpress LLC Supplier No. HYN0368100G
Linalool is a natural monoterpene that acts as a competitive NMDA receptor antagonist. It is orally active and can cross the blood-brain barrier.
  • Anticancer activity
  • Antibacterial properties
  • Anti-inflammatory effects
  • Neuroprotective qualities
  • Anxiolytic effects
  • Antidepressant properties
  • Anti-stress effects
  • Cardioprotective benefits
  • Hepatoprotective effects
  • Nephroprotective effects
  • Pulmonary protective activities