Learn More
Apexbio Technology LLC Linalool 78-70-6 20mg
SDP
Catalog No. 5000094447
Encompass_Preferred
Linalool (CAS 78-70-6) is a naturally occurring monoterpene alcohol predominantly isolated from Cinnamomum camphora (L ) Presl It exhibits modulatory effects on the central nervous system including interactions with neurotransmitter receptors such as GABA sub A /sub contributing to its observed anxiolytic and neuroprotective properties in preclinical studies Linalool is widely investigated for its potential anti-inflammatory antimicrobial and antioxidant activities Due to these diverse bioactivities it is utilized in biomedical research exploring therapeutic strategies for neurological disorders inflammation and microbial infections Store under sealed cool and dry conditions
Catalog No. 50-000-94447 Supplier Apexbio Technology LLC Supplier No. N241820
Linalool (CAS 78-70-6) is a naturally occurring monoterpene alcohol predominantly isolated from Cinnamomum camphora (L ) Presl It exhibits modulatory effects on the central nervous system including interactions with neurotransmitter receptors such as GABA sub A /sub contributing to its observed anxiolytic and neuroprotective properties in preclinical studies Linalool is widely investigated for its potential anti-inflammatory antimicrobial and antioxidant activities Due to these diverse bioactivities it is utilized in biomedical research exploring therapeutic strategies for neurological disorders inflammation and microbial infections Store under sealed cool and dry conditions