Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linalool | 78-70-6 | 97.2% | 154.25 | C10H18O | 10 MG
SDP

Catalog No. 5000295872
Encompass_Preferred

Linalool (Standard) is an analytical reference standard intended for research and analytical applications. It provides defined physicochemical properties and purity to support method development, calibration, and quality control in analytical workflows.

  • Analytical reference standard for linalool
  • Cas number 78-70-6
  • Molecular formula C10H18O
  • Molecular weight 154.25
  • Purity 97.2% (manufacturer listed)
  • Pack size: 10 mg for analytical use
  • For research use only

Catalog No. 50-002-95872 Supplier Medchemexpress LLC Supplier No. HYN0368R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Linalool (Standard) is an analytical reference standard intended for research and analytical applications. It provides defined physicochemical properties and purity to support method development, calibration, and quality control in analytical workflows.

  • Analytical reference standard for linalool
  • Cas number 78-70-6
  • Molecular formula C10H18O
  • Molecular weight 154.25
  • Purity 97.2% (manufacturer listed)
  • Pack size: 10 mg for analytical use
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.