Learn More
Sigma Aldrich Fine Chemicals Biosciences Linalool 97 FCC FG20KG
Catalog No. 502907749
Encompass
Linalool is a monoterepene compound. It is the major component of many essential oils. Anti-inflammatory properties of (A) linalool a naturally occurring enantiomer has been studied. It is also the major constituent of the basil and thyme essential oil and its anti-microbial effect was determined using the agar well diffusion assay. Linalool is reported to have dose-dependent marked sedative effects at the central nervous system (CNS) including hypnotic anticonvulsant and hypothermic properties. Linalool is reported to have a lemon like odor.
Catalog No. 50-290-7749 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W26350820KGK
Linalool is a monoterepene compound. It is the major component of many essential oils. Anti-inflammatory properties of (A) linalool a naturally occurring enantiomer has been studied. It is also the major constituent of the basil and thyme essential oil and its anti-microbial effect was determined using the agar well diffusion assay. Linalool is reported to have dose-dependent marked sedative effects at the central nervous system (CNS) including hypnotic anticonvulsant and hypothermic properties. Linalool is reported to have a lemon like odor.