Learn More
Medchemexpress LLC Linalool 98 stabil 1kg
SDP
Catalog No. 5000669564
Encompass_Preferred
Linalool is a natural monoterpene which is a competitive NMDA receptor antagonist Linalool is orally active and crosses the blood-brain barrier Linalool has anticancer antibacterial anti-inflammatory neuroprotective anxiolytic antidepressant anti-stress cardioprotective hepatoprotective nephroprotective and pulmonary protective activities[1][2][3][4][5]
Catalog No. 50-006-69564 Supplier Medchemexpress LLC Supplier No. 1KGHYN0368
Linalool is a natural monoterpene which is a competitive NMDA receptor antagonist Linalool is orally active and crosses the blood-brain barrier Linalool has anticancer antibacterial anti-inflammatory neuroprotective anxiolytic antidepressant anti-stress cardioprotective hepatoprotective nephroprotective and pulmonary protective activities[1][2][3][4][5]