Learn More
Medchemexpress LLC Linalool-d3 | 1216673-02-7 | 157.27 | 5 MG
SDP
Catalog No. 5000389997
Encompass_Preferred
Linalool-d3 is the deuterium-labeled version of Linalool, a natural monoterpene found in essential oils of coriander. It acts as a competitive antagonist of the N-methyl d-aspartate (NMDA) receptor, exhibiting anti-tumor and anti-cardiotoxicity activities. It also functions as a PPARα ligand, reducing plasma TG levels and altering hepatic transcriptome and plasma metabolome.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89997 Supplier Medchemexpress LLC Supplier No. HYN0368S5MG
Linalool-d3 is the deuterium-labeled version of Linalool, a natural monoterpene found in essential oils of coriander. It acts as a competitive antagonist of the N-methyl d-aspartate (NMDA) receptor, exhibiting anti-tumor and anti-cardiotoxicity activities. It also functions as a PPARα ligand, reducing plasma TG levels and altering hepatic transcriptome and plasma metabolome.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS