Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-Linalool-d3 (vinyl-d3) | 1216673-02-7 | MFCD11109274 | 95.2% | 157.27 g/mol | C10H15D3O | 10 MG
SDP

Catalog No. 5000296054
Encompass_Preferred

Linalool-d3 is a deuterium-labeled analogue of the natural monoterpene linalool, supplied as a stable-isotope reference standard for analytical and research use. It is intended for use as an internal standard and for isotope-tracing experiments in GC-MS or LC-MS workflows, with documented purity, molecular weight, and solubility information available in the manufacturer datasheet and certificates.

  • Deuterium-labeled internal standard for analytical assays.
  • High purity suitable for quantitation and method validation.
  • Soluble in common organic solvents, including DMSO.
  • Provided in a small, laboratory-scale vial for trace-level work.
  • Recommended storage conditions to preserve stability.

Catalog No. 50-002-96054 Supplier Medchemexpress LLC Supplier No. HYN0368S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linalool-d3 is a deuterium-labeled analogue of the natural monoterpene linalool, supplied as a stable-isotope reference standard for analytical and research use. It is intended for use as an internal standard and for isotope-tracing experiments in GC-MS or LC-MS workflows, with documented purity, molecular weight, and solubility information available in the manufacturer datasheet and certificates.

  • Deuterium-labeled internal standard for analytical assays.
  • High purity suitable for quantitation and method validation.
  • Soluble in common organic solvents, including DMSO.
  • Provided in a small, laboratory-scale vial for trace-level work.
  • Recommended storage conditions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.