Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linalool oxide | 60047-17-8 | 98.0% | 170.25 | 1 G
SDP

Catalog No. 5000426333
Encompass_Preferred

Linalool oxide is a secondary metabolite found in elongating wheat plants. It is known for its antinociceptive and anticonvulsant effects, and it also exhibits anxiolytic activity. This product is intended for research use only.

  • Purity of 98.0%
  • Molecular weight of 170.25
  • Chemical formula C10H18O2
  • CAS number 60047-17-8
  • Appears as a colorless to light yellow liquid
  • Exhibits antinociceptive and anticonvulsant effects
  • Shows anxiolytic activity

Catalog No. 50-004-26333 Supplier Medchemexpress LLC Supplier No. HYN95021G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linalool oxide is a secondary metabolite found in elongating wheat plants. It is known for its antinociceptive and anticonvulsant effects, and it also exhibits anxiolytic activity. This product is intended for research use only.

  • Purity of 98.0%
  • Molecular weight of 170.25
  • Chemical formula C10H18O2
  • CAS number 60047-17-8
  • Appears as a colorless to light yellow liquid
  • Exhibits antinociceptive and anticonvulsant effects
  • Shows anxiolytic activity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.