Learn More
Medchemexpress LLC Linalyl acetate | 115-95-7 | 1 G
SDP
Catalog No. 5000419155
Encompass_Preferred
Linalyl acetate is a primary component found in many plant essential oils, demonstrating a range of biological activities including anti-anxiety, anti-inflammatory, anti-diabetic, anti-stress, and cardiovascular-regulatory effects. It is orally active and suitable for research purposes.
  • Demonstrates anti-anxiety and sedative effects
  • Exhibits cardiovascular-regulatory properties
  • Provides anti-inflammatory and anti-fibrotic benefits
  • Helps prevent mitochondrial dysfunction
  • Regulates inflammatory mediators
Catalog No. 50-004-19155 Supplier Medchemexpress LLC Supplier No. HYN69481G
Linalyl acetate is a primary component found in many plant essential oils, demonstrating a range of biological activities including anti-anxiety, anti-inflammatory, anti-diabetic, anti-stress, and cardiovascular-regulatory effects. It is orally active and suitable for research purposes.
  • Demonstrates anti-anxiety and sedative effects
  • Exhibits cardiovascular-regulatory properties
  • Provides anti-inflammatory and anti-fibrotic benefits
  • Helps prevent mitochondrial dysfunction
  • Regulates inflammatory mediators