Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linalyl acetate | 115-95-7 | MFCD00008907 | 100 MG
SDP

Catalog No. 5000417960
Encompass_Preferred

Linalyl acetate is a principal component of many plant essential oils, known for its anti-anxiety, anti-inflammatory, anti-diabetic, anti-stress, and cardiovascular-regulatory effects. It is orally active and presents as a colorless to light yellow liquid with a purity of 98.80% and a density of 0.8997 g/cm³.

  • Molecular weight: 196.29
  • Chemical formula: C12H20O2
  • Exhibits sedative and anti-anxiety effects
  • Reduces heart rate and LDH activity
  • Alleviates cardiovascular and cell damage from acute nicotine exposure
  • Regulates inflammatory mediators and prevents mitochondrial dysfunction
  • Exhibits anti-inflammatory and anti-fibrotic effects

Catalog No. 50-004-17960 Supplier Medchemexpress LLC Supplier No. HYN6948100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linalyl acetate is a principal component of many plant essential oils, known for its anti-anxiety, anti-inflammatory, anti-diabetic, anti-stress, and cardiovascular-regulatory effects. It is orally active and presents as a colorless to light yellow liquid with a purity of 98.80% and a density of 0.8997 g/cm³.

  • Molecular weight: 196.29
  • Chemical formula: C12H20O2
  • Exhibits sedative and anti-anxiety effects
  • Reduces heart rate and LDH activity
  • Alleviates cardiovascular and cell damage from acute nicotine exposure
  • Regulates inflammatory mediators and prevents mitochondrial dysfunction
  • Exhibits anti-inflammatory and anti-fibrotic effects

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.