Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linalyl acetate, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019818 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9818 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9818 Supplier MilliporeSigma Supelco Supplier No. 49599100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified analytical standard suitable for chromatographic applications including HPLC and gas chromatography.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Linalyl Acetate
CAS 115-95-7
Vapor Pressure 0.1 mmHg (20°C)
Vapor Density 6.8 (vs. air)
Density 1.448-1.452 at 20°C; 0.901 g/mL (at 25°C)
Flash Point 85°C
Physical Form Neat
Linear Formula CH3 CO2 C(CH=CH2)(CH3)CH2 CH2 CH=C(CH3)2
Technique HPLC
MDL Number MFCD00008907
Boiling Point 220°C
Synonym 3,7-Dimethyl-1,6-octadien-3-yl acetate; Bergamol
Type Analytical Standard
Solvent or Matrix Neat
Molecular Weight (g/mol) 196.29
Industry Type Flavors and Fragrances
Quantity 100 mg
Formula Weight 196.29
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.