Learn More
Sigma Aldrich Fine Chemicals Biosciences Linalyl acetate natural 961KG
Catalog No. 502902400
Encompass
Linalyl acetate is a monoterpene ester mainly found in lavandula essential oil. It is used as a flavoring agent in food industries as well as a preservative additive in cosmetics and fragrances such as soaps colognes perfumes and skin lotions.
Catalog No. 50-290-2400 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W2636131KGK
Linalyl acetate is a monoterpene ester mainly found in lavandula essential oil. It is used as a flavoring agent in food industries as well as a preservative additive in cosmetics and fragrances such as soaps colognes perfumes and skin lotions.