Learn More
Medchemexpress LLC Linaprazan | 248919-64-4 | MFCD30343852 | 98.8% | 366.46 g/mol | C21H26N4O2 | 25 MG
SDP
Catalog No. 5000214787
Encompass_Preferred
Linaprazan is a potassium-competitive H+,K+-ATPase inhibitor supplied for research and analytical applications. It exhibits K+-competitive inhibition of the gastric proton pump (reported IC50 ≈ 1.0 ± 0.2 μM) and has demonstrated oral activity in preclinical studies of acid-related disorders. This material is for research use only and not intended for human use.
  • Potassium-competitive H+,K+-ATPase inhibition (IC50 ≈ 1.0 μM).
  • Orally active in preclinical models of reflux and acid suppression.
  • Available as solid and DMSO solution formats.
  • High purity suitable for analytical and research applications.
  • Multiple package sizes available, including 25 mg.
Catalog No. 50-002-14787 Supplier Medchemexpress LLC Supplier No. HY10041225MG
Linaprazan is a potassium-competitive H+,K+-ATPase inhibitor supplied for research and analytical applications. It exhibits K+-competitive inhibition of the gastric proton pump (reported IC50 ≈ 1.0 ± 0.2 μM) and has demonstrated oral activity in preclinical studies of acid-related disorders. This material is for research use only and not intended for human use.
  • Potassium-competitive H+,K+-ATPase inhibition (IC50 ≈ 1.0 μM).
  • Orally active in preclinical models of reflux and acid suppression.
  • Available as solid and DMSO solution formats.
  • High purity suitable for analytical and research applications.
  • Multiple package sizes available, including 25 mg.