Learn More
Medchemexpress LLC Linaprazan glurate | 1228559-81-6 | 99.7% | C26H32N4O5 | 100 MG
SDP
Catalog No. 5000356199
Encompass_Preferred
Linaprazan glurate (X842) is an orally active prodrug of Linaprazan that provides a potent and prolonged inhibitory effect on gastric acid secretion. It is rapidly transformed by enzymatic cleavage into its active metabolite, linaprazan. It acts as a potassium-competitive acid blocker, selectively inhibiting acid formation from gastric H⁺/K⁺-ATPase in a potassium-dependent manner. This product can be utilized for studies related to erosive esophagitis (EE).
  • Orally active prodrug
  • Potent and prolonged inhibitory effect on gastric acid secretion
  • Rapidly transformed into active metabolite
  • Potassium-competitive acid blocker
  • Selectively inhibits acid formation from gastric H⁺/K⁺-ATPase
  • Can be used for studies of erosive esophagitis (EE)
  • Appearance: solid
  • Color: white to off-white
Catalog No. 50-003-56199 Supplier Medchemexpress LLC Supplier No. HY145578100MG
Linaprazan glurate (X842) is an orally active prodrug of Linaprazan that provides a potent and prolonged inhibitory effect on gastric acid secretion. It is rapidly transformed by enzymatic cleavage into its active metabolite, linaprazan. It acts as a potassium-competitive acid blocker, selectively inhibiting acid formation from gastric H⁺/K⁺-ATPase in a potassium-dependent manner. This product can be utilized for studies related to erosive esophagitis (EE).
  • Orally active prodrug
  • Potent and prolonged inhibitory effect on gastric acid secretion
  • Rapidly transformed into active metabolite
  • Potassium-competitive acid blocker
  • Selectively inhibits acid formation from gastric H⁺/K⁺-ATPase
  • Can be used for studies of erosive esophagitis (EE)
  • Appearance: solid
  • Color: white to off-white