Learn More
Medchemexpress LLC Linaprazan glurate | 1228559-81-6 | 99.7% | C26H32N4O5 | 10MG
SDP
Catalog No. 5000258600
Encompass_Preferred
Linaprazan glurate is a small-molecule research compound that acts as a potassium-competitive acid blocker and is used in preclinical studies of gastric acid secretion and proton pump-related mechanisms. It is supplied as a high-purity solid for analytical, synthetic, and medicinal chemistry applications.
Catalog No. 50-002-58600 Supplier Medchemexpress LLC Supplier No. HY14557810MG
Linaprazan glurate is a small-molecule research compound that acts as a potassium-competitive acid blocker and is used in preclinical studies of gastric acid secretion and proton pump-related mechanisms. It is supplied as a high-purity solid for analytical, synthetic, and medicinal chemistry applications.