Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Linarin (buddleoside) | 480-36-4 | MFCD00151178 | 98.0% | 592.55 | C28H32O14 | 10 MG
SDP

Catalog No. 5000296139
Encompass_Preferred

Linarin (buddleoside) is a naturally occurring flavonoid glycoside and an orally active, selective acetylcholinesterase (AChE) inhibitor. It has reported anti-inflammatory, antioxidant, sedative, bone differentiation, antitumor, antibacterial, and antiviral activities and is used in research on nervous system disorders, osteoporosis, and cancer.

  • Natural flavonoid glycoside, also known as buddleoside.
  • Selective acetylcholinesterase (AChE) inhibitor, orally active.
  • CAS 480-36-4; formula C28H32O14; molecular weight 592.55.
  • Purity typically 98.0% as supplied.
  • White to off-white solid; soluble in DMSO (≈250 mg/mL).
  • Storage: protect from light; short-term 4°C; in solution -20°C to -80°C as specified.
  • Supplied in small research quantities suitable for in vitro and in vivo studies.

Catalog No. 50-002-96139 Supplier Medchemexpress LLC Supplier No. HYN052810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Linarin (buddleoside) is a naturally occurring flavonoid glycoside and an orally active, selective acetylcholinesterase (AChE) inhibitor. It has reported anti-inflammatory, antioxidant, sedative, bone differentiation, antitumor, antibacterial, and antiviral activities and is used in research on nervous system disorders, osteoporosis, and cancer.

  • Natural flavonoid glycoside, also known as buddleoside.
  • Selective acetylcholinesterase (AChE) inhibitor, orally active.
  • CAS 480-36-4; formula C28H32O14; molecular weight 592.55.
  • Purity typically 98.0% as supplied.
  • White to off-white solid; soluble in DMSO (≈250 mg/mL).
  • Storage: protect from light; short-term 4°C; in solution -20°C to -80°C as specified.
  • Supplied in small research quantities suitable for in vitro and in vivo studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.