Learn More
Cayman Chemical LInarmycIn A 2.5mg
SDP
Catalog No. 502947892
Encompass_Preferred
A polyene antibiotic; active against S. aureus, E. coli, S. cerevisiae, C. albicans, and A. niger in disc assays (MICs = 3.1, 1.6, 0.1, 1.6, and 0.2 UG/disc, respectively); induces lysis and degradation of B. subtilis as a component of Streptomyces Mg1 extract
Catalog No. 50-294-7892 Supplier Cayman Chemical Supplier No. 331322.5 MG
A polyene antibiotic; active against S. aureus, E. coli, S. cerevisiae, C. albicans, and A. niger in disc assays (MICs = 3.1, 1.6, 0.1, 1.6, and 0.2 UG/disc, respectively); induces lysis and degradation of B. subtilis as a component of Streptomyces Mg1 extract