Learn More
MP Biomedicals™ Linbro™ Liquisystem, Cap-Bands
Catalog No. ICN61227B1 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
120/Cs.
Catalog No. Quantity
ICN61227B1 120/Cs.
1 options
Catalog No. ICN61227B1 Supplier MP Biomedicals™ Supplier No. 0961227B1
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Used with combination of Tube 96 & I.D. Labels 96 to form a complete system for the storage of samples up to 1.0 mL is created. Once filled with samples, the tube can be sealed in an air tight fashion using the Cap Band 8.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Linbro Liquisystem
For Use With (Application) Used with combination of Tube 96 and I.D. Labels 96 to form a complete system for the storage of samples up to 1mL is created.
Quantity 120/Cs.