Learn More
MSC Linco Swivel Top Plate Caster, Polyurethane, 6 in Wheel Dia, 3,600 lb Capacity, Side Lock Brake
Catalog No. 5000529504
Encompass
This swivel top plate caster features a polyurethane wheel with a 6 in diameter and 2 in width. It offers a 3,600 lb capacity and an overall height of 7-1/2 in. The caster includes a side lock brake for securing its position, and is constructed from heavy gauge steel with a zinc finish. It utilizes a ball bearing type and has a 4 x 4-1/2 in mounting plate.
  • Polyurethane wheel material
  • 6 in wheel diameter
  • 2 in wheel width
  • 3,600 lb load capacity
  • 7-1/2 in overall height
  • Side lock brake
  • Heavy gauge steel construction
  • Zinc finish/coating
  • Ball bearing type
  • 4 x 4-1/2 in plate size
  • Red wheel color
  • Swivel mount type
Catalog No. 50-005-29504 Supplier MSC Supplier No. 12050811
This swivel top plate caster features a polyurethane wheel with a 6 in diameter and 2 in width. It offers a 3,600 lb capacity and an overall height of 7-1/2 in. The caster includes a side lock brake for securing its position, and is constructed from heavy gauge steel with a zinc finish. It utilizes a ball bearing type and has a 4 x 4-1/2 in mounting plate.
  • Polyurethane wheel material
  • 6 in wheel diameter
  • 2 in wheel width
  • 3,600 lb load capacity
  • 7-1/2 in overall height
  • Side lock brake
  • Heavy gauge steel construction
  • Zinc finish/coating
  • Ball bearing type
  • 4 x 4-1/2 in plate size
  • Red wheel color
  • Swivel mount type