Learn More
Med Vet International Lincobac 300 (Lincomycin) Injection, Swine Antibiotic, 100mL, Each
Catalog No. 502828908
Encompass_Preferred
LincoBac 300 is indicated for the treatment of infectious forms of arthritis in swine caused by organisms including staphylococci, streptococci, Erysipelothrix and Mycoplasma spp. It is also indicated for the treatment of mycoplasma pneumonia. The injection contains the proven antibiotic lincomycin. Use in all ages, at all stages of production. Contains 300 mg/mL.
Catalog No. 50-282-8908 Supplier Med Vet International Supplier No. RXLINCO300100INJV
LincoBac 300 is indicated for the treatment of infectious forms of arthritis in swine caused by organisms including staphylococci, streptococci, Erysipelothrix and Mycoplasma spp. It is also indicated for the treatment of mycoplasma pneumonia. The injection contains the proven antibiotic lincomycin. Use in all ages, at all stages of production. Contains 300 mg/mL.