Learn More
MSC Lincoln Electric VIKING 1740 Ignition, Auto-Darkening, Shade 9-13, 1 Each
Catalog No. 5000769317
Encompass
The VIKING 1740 series welding helmet offers superior optics, control, and comfort. Featuring 4C® Lens Technology and 1/1/1/1 optical clarity, it enhances the visible color spectrum, eliminates imperfections, and provides variable 9-13 shades. It includes a grind mode, LED safety indicator, auto on/off, solar power, and replaceable AAA batteries. Its lightweight design and adjustable ratchet headgear ensure all-day comfort.
  • Proprietary 4C® Lens Technology enhances visible color spectrum for clearer viewing.
  • Achieves 1/1/1/1 optical clarity, eliminating distortion and providing consistent shade.
  • Features a 6.3 sq. inch (3.78 x 1.67 in.) auto-darkening viewing area.
  • Variable 9-13 shade with delay and sensitivity controls.
  • Grind mode with LED safety indicator.
  • Lightweight 18.6 oz. (528 g) design.
  • Equipped with 3-point pivot headgear for weight distribution.
Catalog No. 50-007-69317 Supplier MSC Supplier No. 29331220
The VIKING 1740 series welding helmet offers superior optics, control, and comfort. Featuring 4C® Lens Technology and 1/1/1/1 optical clarity, it enhances the visible color spectrum, eliminates imperfections, and provides variable 9-13 shades. It includes a grind mode, LED safety indicator, auto on/off, solar power, and replaceable AAA batteries. Its lightweight design and adjustable ratchet headgear ensure all-day comfort.
  • Proprietary 4C® Lens Technology enhances visible color spectrum for clearer viewing.
  • Achieves 1/1/1/1 optical clarity, eliminating distortion and providing consistent shade.
  • Features a 6.3 sq. inch (3.78 x 1.67 in.) auto-darkening viewing area.
  • Variable 9-13 shade with delay and sensitivity controls.
  • Grind mode with LED safety indicator.
  • Lightweight 18.6 oz. (528 g) design.
  • Equipped with 3-point pivot headgear for weight distribution.