Learn More
Medchemexpress LLC Lincomycin-d3 | 154-21-2 | 99.9% | 409.56 | 500 UG
SDP
Catalog No. 5000436208
Encompass_Preferred
Lincomycin-d3 is the deuterium labeled Lincomycin, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Deuteration may affect pharmacokinetic and metabolic profiles
  • Solid appearance
  • White to off-white color
  • Protect from light
  • Store under nitrogen
Catalog No. 50-004-36208 Supplier Medchemexpress LLC Supplier No. HY117660S500UG
Lincomycin-d3 is the deuterium labeled Lincomycin, intended for research use only. This compound serves as a tracer and an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Deuteration may affect pharmacokinetic and metabolic profiles
  • Solid appearance
  • White to off-white color
  • Protect from light
  • Store under nitrogen