Learn More
Medchemexpress LLC Lincomycin-d3 | 154-21-2 | 99.90% | 409.56 | 1 MG
SDP
Catalog No. 5000437151
Encompass_Preferred
Lincomycin-d3 is the deuterium labeled Lincomycin. This compound is used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated for quantitation during drug development
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-37151 Supplier Medchemexpress LLC Supplier No. HY117660S1MG
Lincomycin-d3 is the deuterium labeled Lincomycin. This compound is used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during the drug development process. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated for quantitation during drug development
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs