Learn More
Apexbio Technology LLC Lincomycin HCl 859-18-7 5g
SDP
Catalog No. 5000093103
Encompass_Preferred
Lincomycin HCl (859-18-7) is a small-molecule inhibitor targeting bacterial protein synthesis It is designed to inhibit translation by binding to the 50S subunit of the bacterial ribosome thereby blocking peptide elongation Lincomycin HCl exerts its biological activity primarily through inhibition of bacterial protein synthesis In in vitro studies Lincomycin HCl demonstrates bacteriostatic activity with reported IC50 values ranging from approximately 0 5 g/mL to 2 g/mL depending on bacterial strain and culture conditions Based on these pharmacological properties Lincomycin HCl holds research potential in selective antibiotic assays bacterial resistance studies and maintenance of cell culture sterility
Catalog No. 50-000-93103 Supplier Apexbio Technology LLC Supplier No. B19625000
Lincomycin HCl (859-18-7) is a small-molecule inhibitor targeting bacterial protein synthesis It is designed to inhibit translation by binding to the 50S subunit of the bacterial ribosome thereby blocking peptide elongation Lincomycin HCl exerts its biological activity primarily through inhibition of bacterial protein synthesis In in vitro studies Lincomycin HCl demonstrates bacteriostatic activity with reported IC50 values ranging from approximately 0 5 g/mL to 2 g/mL depending on bacterial strain and culture conditions Based on these pharmacological properties Lincomycin HCl holds research potential in selective antibiotic assays bacterial resistance studies and maintenance of cell culture sterility